Learn More
Proteintech™ HumanKine™ Recombinant Human G-CSF Protein
Tous les produits Proteintech Produits
Click to view available options
Quantité:
10 μg
100 μg
1 000 μg
Conditionnement:
1000µg
100µg
10µg
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spécification
Spécification
| Identification génétique (Entrez) | 1440 |
| Poids moléculaire | 21 to 25 kDa, monomer, glycosylated |
| Quantité | 100 μg |
| Espèces | Human |
| Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
| Contenu et stockage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| Activité | Typically > 0.1 ng/mL EC50 |
| Système d’expression | Human embryonic kidney 293 (HEK293) human cell expression system |
| Forme | Lyophilized |
| Pureté ou qualité | > 95% |