Learn More
Proteintech™ HumanKine™ Recombinant Human HSA Protein
Code produit. 16851426 Tous les produits Proteintech Produits
Click to view available options
Quantité:
10 x 1 000 μg
100 μg
1 000 μg
Conditionnement:
10 pièces
1000µg
100µg
Code produit. 16851426
Marque: Proteintech™ HZ30011000UG
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spécification
Identification génétique (Entrez) 213
Poids moléculaire 60 to 65 kDa, monomer, glycosylated
Quantité 1 000 μg
Espèces Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS.
Contenu et stockage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Système d’expression Human embryonic kidney 293 (HEK293) human cell expression system
Forme Lyophilized
Pureté ou qualité > 95%