Learn More
Ambion™ mirVana™ miRNA Inhibitor
Description
mirVana miRNA Inhibitors are single-stranded RNA molecules designed to inhibit endogenous microRNAs (miRNAs) and are ideal for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications. Based on the latest version of miRbase, mirVana miRNA Inhibitors are available for a wide range of miRNAs, providing researchers with a powerful tool to investigate gene regulation mechanisms.
Spécification
Spécification
| Contenu et stockage | Conserver au congélateur (-5 à -30°C). |
| Description | Inhibiteurs de micro-ARN mirVana™, 5 nmol, ARN à simple brin de petite taille, modifié chimiquement, inhibition de l’ARN endogène, régulation négative de l’activité des micro-ARN |
| Quantité | 5 nmol |
Usage exclusivement réservé à la recherche.Ne pas utiliser pour des procédures de diagnostic.