Learn More
Brady™ i75 Series R6700 Ribbon for the i7500 Label Printer
Click to view available options
Alternate Part Number:
i75-R6700-WT
i75-R6702-WT
i75-R6707-WT
i75-R7962-110X300
Conditionnement:
1 pièce
Description
- i75-series ribbon is self-centering with ink-side always correctly facing the label
- One-color ribbon for printing a single color onto a label
- R6700 Series White Wax/Resin formulation offers excellent chemical, scratch and smudge resistance when printed on the appropriate label material from Brady™
- Recommended for printing on dark-colored heat-shrinkable tubing materials, such as black B-342 and B-344 PermaSleeve™ wire markers from Brady™
- Combine Brady label materials with their recommended ribbon series for superior legend performance
Spécification
Spécification
| Type de produit | Printer Ribbon |
| Couleur | White |
| À utiliser avec (équipement) | i7500 |
| Matériau | Polyester |
| Print Technology | Ink Jet |
| Longueur (impériale) | 328.08 yd. |
| Largeur (impériale) | 2.36 in. |
| Longueur (métrique) | 300 m |
| Largeur (métrique) | 60 mm |
| Épaisseur (métrique) | 4.5 μm |
| Afficher plus |