Learn More
IgM (H+L) Mouse anti-Rabbit, PerCP, Novus Biologicals™
Code produit 18039105
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18039105 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18039105 Fournisseur Bio-Techne Code fournisseur NBP268467PCP
Mouse Monoclonal Antibody
IgM (H+L) Monoclonal antibody specifically detects IgM (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Spécification
Antigène IgM (H+L)
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone NRBM
Conjugué PerCP
Formule PBS
Espèces hôtes Mouse
Immunogène Rabbit IgM
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche B Cell Development and Differentiation Markers, Immunology
Primaire ou secondaire Secondary
Espèces cibles Rabbit
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG1