Learn More
Tocris Bioscience™ Indo 1AM
Click to view available options
Quantité:
1 mg
Conditionnement:
1mg
Description
Indo 1AM is a fluorescent Ca2+ indicator (Kd for Ca2+ = 250 nM). Cell-permeable. Displays ratiometric emission wavelength change upon Ca2+ binding. Excitation/emission λ are 355/475 nm for no Ca2+ and 355/401 nm for high Ca2+. Absorption maxima are 349 and 331 nm for free anion and Ca complex, respectively. Respective emission maxima are 485 and 410 nm. Quantum yield ~0.5. Ideal for flow cytometry.
Spécification
Spécification
| Nom chimique ou matériau | 2-[4-[Bis[2-[(acetoxy)methoxy]-2-oxoethyl]amino]-3-[2-[2-[bis[2-[(acetyloxy)methoxy]-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]phenyl]-1H-indole-6-carboxylic acid (acetoxy)methyl ester |
| CAS | 112926-02-0 |
| Quantité | 1 mg |
| Cible | Calcium Signaling Probes and Ca2+ Indicator Dyes |
| Formule moléculaire | C47H51N3O22 |
| Poids de la formule | 1009.91 |
| Pureté | 90% |