Learn More
Invitrogen™ PCR SuperMix High Fidelity
Description
Includes
PCR SuperMix High Fidelity is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 100 amplification reactions of 50μL each are provided. PCR SuperMix High Fidelity Components: A 22U/mL DNA polymerase mixture in 66mM Tris-SO4 (pH 9.1 at 25°C), 19.8mM (NH4)2SO4, 2.2mM MgSO4, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP and stabilizers. 100 rxns: 4 x 1.125mL.
Requires
Matrices et amorces
• Pour éviter le mispriming
• PCR
• PCR SuperMix High Fidelity
• Composition:, , 66mm Tris-SO4 (pH 9,1 25°C), , 19,8mm (NH4)2SO4, , 11mm KCl, , 2,2mm MgSO4, , 220 μM dNTPs, , stabilisants
Informations sur la commande
Produit homologué pour des expéditions sur de la glace carbonique ou mouillée
Spécification
Spécification
| Quantité | 100 reactions |
| Comprend | PCR SuperMix High Fidelity is supplied at 1.1X concentration to allow approximately 10% of the final reaction volume to be used for the addition of primer and template solutions. Reagents sufficient for 100 amplification reactions of 50μL each are provided. PCR SuperMix High Fidelity Components: A 22U/mL DNA polymerase mixture in 66mM Tris-SO4 (pH 9.1 at 25°C), 19.8mM(NH4)2SO4, 2.2mM MgSO4, 220μM dGTP, 220μM dATP, 220μM dTTP, 220μM dCTP and stabilizers. 100 rxns: 4 x 1.125mL. Store at -20°C in a non-frost-free freezer. |