Learn More
Astell Scientific™ IQ/OQ Documentation Service
Validate your autoclave with installation qualification and operational qualification. Astell Scientific™ IQ/OQ Documentation Service ensures that your desired sterilization outcome is always met.
Marque:  Astell Scientific™ IQ/OQ001
Code produit. 15304895
  • 1330.00€ / 1 pièce
Il en reste null
Expédition estimée: 06-07-2026
Description
  • Validation of a new steam sterilizer is required to verify that the unit is capable of delivering the tasks and performing to the parameters set out in the original URS.
  • In the heavily regulated pharmaceutical industry, accuracy and repeatability are imperative to both research and production.
  • Validating an autoclave with IQ/OQ/PQ benchmarks the installation, operation and performance of the autoclave from the design specifications and manufacturer recommendations.
  • The IQ/OQ option can be specified on any Astell™ circular or square section autoclave.
  • The IQ documentation includes, as a minimum, details of calibration equipment, an order acknowledgement, PED (pressure equipment directive) compliance, a declaration of conformity, FAT (factory acceptance test), a drawing schedule, ISO 9001:2015 and ISO 13485:2012 certification, pressure vessel specification and door safety checks.
  • The OQ documentation includes an automatic control test sheet (per cycle) and chamber temperature distribution (per cycle).
  • Specifying IQ/OQ with your next autoclave will give you peace of mind, as it establishes documented evidence that shows a high degree of assurance that the sterilization process will be performed consistently.
  • IQ/OQ can be especially valuable where a custom built autoclave is requested. Astell™ has the experience and knowledge to help a customer spec. the equipment they need and write protocols to exact customer specifications. For more information please contact us.
Spécification
Spécification
ISO 9001:2015 and ISO 13485:2012
Any Astell™ circular or square section autoclave