Learn More
Isobutyramide, 99 %, Thermo Scientific Chemicals
Code produit. 11460197
Click to view available options
Quantité:
25 g
100 g
500 g
Conditionnement:
100g
25g
500g
Code produit. 11460197
Marque: Thermo Scientific Alfa Aesar B21641.14
Demander un format en vrac ou personnalisé
CAS: 563-83-7 | C4H9NO | 87.122 g/mol
Isobutyramide plays an important role in chemical grafting of human serum albumin during the synthesis of sequentially assembled protein capsules. It is used in pharmaceutical manufacturing and in cellular differentiation. It finds application in the transcription of human gamma-globin gene and murine embryonic epsilon(y)-globin gene. Further, it is in the treatment of beta-thalassemia and sickle cell disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubilité
Soluble dans l’eau.
Remarques
Incompatible avec les acides, les bases, les agents oxydants forts et les agents réducteurs forts.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 563-83-7
Formule moléculaire C4H9NO
Poids moléculaire (g/mol) 87.122
Numéro MDL MFCD00008019
Clé InChI WFKAJVHLWXSISD-UHFFFAOYSA-N
Synonyme isobutyramide, propanamide, 2-methyl, 2-methylpropionamide, isobutyrimidic acid, isobutylamide, isopropylformamide, dimethylacetoamide, unii-82uoe7b38z, dimethylacetamid, c-isopropylformamide
CID PubChem 68424
Nom IUPAC 2-méthylpropanamide
SMILES CC(C)C(=O)N
Spécification
Nom chimique ou matériau Isobutyramide
Point de fusion 127°C to 129°C
Densité 1.013
Point d’ébullition 216°C to 220°C
Quantité 25 g
Informations sur la solubilité Soluble in water.
Poids de la formule 87.12
Pourcentage de pureté 99%