Learn More
Novus Biologicals™ Kidney Matched Tumor and Normal Tissue Lysate
Code produit 18120188 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
2 flacons
Conditionnement:
2 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18120188 2 flacons 2 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18120188 Fournisseur Novus Biologicals™ Code fournisseur NBP247100
  • ONLINE EXCLUSIVE PRICE
    496.00€
    468.30€ / 2 pièces
    Économisez 27.70€
    5% de réduction
 Article restreint
Expédition estimée: 04-11-2026
Two matched 1mg/mL vials (100μg/vial) of human kidney tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Kidney Matched Tumor & Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Spécification
Tissue Kidney
Quantité 2 flacons
Espèces hôtes Human
Contenu et stockage Store at −80°C. Avoid freeze-thaw cycles.