Learn More
LNI Swissgas Kit service for NG SIRIO 5000 HP
Code produit. 17862019 Tous les produits LNI Swissgas Produits
Click to view available options
Type:
Kit PM
Conditionnement:
1 pièce
Code produit. 17862019
Marque: LNI Swissgas 6930.00.190
Kit service 8000 h for NG SIRIO 5000 HP.
  • To be carried out by trained service engineer every 16000 hours or 4 years.
Spécification
Type Kit PM