missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TerraCycle™ Laboratory PPE and Plastic Recycling Zero Waste Box™

Click to view available options
Dimensions:
Petit
Moyen
Grand
Conditionnement:
1 pièce
Description
Use this Zero Waste Box™ to recycle:
- Disposable PPE (face masks, gloves, garments, beard nets, disposable shoe covers, earplugs, hair nets)
- Protective eyewear (safety glasses)
- Rigid lab plastics
- Flexible lab plastics
- Disinfectant wipes
- We accept all types of disposable face masks made from plastic, such as dust masks, FFP2 (or so-called 'N95'), FFP3 and surgical masks.
- Do not send in:
- Medical waste. This includes material contaminated with blood, bodily fluids and waste, which should be collected in a hazardous clinical waste bin
- Lab plastics that are contaminated with hazardous waste
- Fabric, homemade or paper face masks
- Wax earplugs
- Plant-based and biogradable plastics and wipes
- Autoclaved material
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic)
- Batteries, pressurised canisters, broken glass
Spécification
Spécification
| Capacité (métrique) | 78 L |
| Capacité (impériale) | 2.8 cu.ft. |
| Couleur | Beige, Green, Red, White |
| Dimensions (L x l x H) | 30.5 x 30.5 x 84 cm |
| Jetable | Single-use |
| Sans latex | Yes |
| Dimensions | Moyen |
| Stérilité | Non-sterile |
| Autoclavable | Not Autoclavable |
| Matériau | Cardboard, Plastic |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu