Learn More
Lambda-Cyhalothrin, TRC
Code produit 30137701
Change view
Click to view available options
Quantité:
10 mg
100 mg
Conditionnement:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30137701 100 mg 100mg
18083350 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30137701 Fournisseur Toronto Research Chemicals Code fournisseur TRCC988980100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 91465-08-6
Formule moléculaire C23 H19 Cl F3 N O3
Poids moléculaire (g/mol) 449.85
Synonyme Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1α(S*),3α(Z)]-(±)-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (R)-cyano(3-phenoxyphenyl)methyl ester, (1S,3S)-rel- (9CI), A 12871A, Ambush Super, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (1R)-cyano(3-phenoxyphenyl)methyl ester, (1S,3S)-, Cyhalothrin K, Demand, Demand Pestab, Grenade ER, Hallmark, ICI-A 0321, ICON Maxx, Icon, Karate, Karate 1E, Karate 2.5EC, Karate K, Karate King, Karate Z, Karate Zeon, Karate Zeon 050CS, Karate Zeon 100CS, Kung Fu, Lamdex, Matador, Matador (insecticide), Molniya, NUP 05077, PP 321, REEVA 5, Scimitar, Spectracide Trazicide, Spectracide Triazicide Soil & Turf Insect Killer, Taiga Z, Warrior, Warrior II, Warrior T, Warrior Zeon, lambda-Cyhalothrin, λ-Cyhalothrin
Nom IUPAC [(R)-cyano-(3-phenoxyphenyl)methyl] (1S,3S)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C)[C@H](\C=C(/Cl)\C(F)(F)F)[C@@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2
Spécification
Nom chimique ou matériau lambda-Cyhalothrin
Noms d’analytes ou de composants lambda-Cyhalothrin
Notes de qualité de la pureté HPLC
Conditions d’expédition Room Temperature
Formule InChI InChI=1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/b19-12-/t17-,18+,20-/m1/s1
Type de solution Neat
Industry Type Pharmaceutical
Quantité 100 mg
Poids de la formule 449.1
Pourcentage de pureté >95%