Learn More
Gilac™ Large Volume Container with Pivoting Wheels and Brakes
Click to view available options
Capacité (métrique):
100 l
170 l
220 l
310 l
500 l
Comprend:
2 roues pivotantes avec freins
4 roues pivotantes, 2 freins
Conditionnement:
1 pièce
Description
- The economical solution for moving large volume containers around.
- Stainless steel wheels and fastenings.
- Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
- Load capacity for information purposes only (depending on how the load is spread and the operating temperature).
- Bottom with gaskets on the axle for perfect sealing.
- Volume in litres is equivalent to the container filled to the brim.
Spécification
Spécification
| Type | Storage Tote |
| Matériau | HDPE |
| Couleur | Blanc |
| Forme | Rectangulaires |
| Capacité (impériale) | 44.9 gal. |
| Capacité (métrique) | 170 l |
| Comprend | 2 roues pivotantes avec freins |