Learn More
Leupeptin, MP Biomedicals™
Code produit. 11438191 Tous les produits MP Biomedicals Produits
Click to view available options
Quantité:
5 mg
25 mg
100 mg
Conditionnement:
100mg
25mg
5mg
Code produit. 11438191
Marque: MP Biomedicals 0215155305
Inhibits trypsin-like proteases and some cysteine proteases. MP Biomedicals' Leupeptin is a modified tripeptide reversible protease inhibitor. Little to no inhibition is seen against pepsin, cathepsins A and D, and alpha-chymotrypsin.
When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.
Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
TRUSTED_SUSTAINABILITY
Spécification
CAS 103476-89-7
Contenu et stockage Store at −20°C, desiccate, protect from light.
Format White Powder
Poids moléculaire 427.57
Quantité 5 mg