Learn More
Brady Magicard™ Prima 8 Duo
Click to view available options
Couleur:
Blanc, Bleu
Conditionnement:
1 pièce
Description
- Print high-quality full-color cards over the edge on both sides
- Equipped with a range of card encoding devices to write secure electronic data to cards at the point of issuance
- 600 dpi printing for ultra-sharp images
Specifications
Specifications
| Type de produit | Printer |
| Couleur | Blanc, Bleu |
| Type d’affichage | Monocolor LCD |
| À utiliser avec (équipement) | Brady Labels |
| Matériau | Plastic |
| Print Technology | Thermal Transfer |
| Longueur (impériale) | 18.7 in. |
| Largeur (impériale) | 8.3 in. |
| Longueur (métrique) | 475.5 mm |
| Largeur (métrique) | 211.5 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction