Learn More
Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents
Tous les produits Applied Biosystems ProduitsDescription
The MagMAX Sequential DNA/RNA kit and reagents, when paired with a KingFisher purification system, are an automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples. This innovative kit and standalone reagents eliminate the need to purchase and run separate DNA and RNA purification kits, streamlining your workflow and maximizing efficiency.
The MagMAX Sequential DNA/RNA kit and reagents, when paired with a KingFisher purification system, are an automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples. This innovative kit and standalone reagents eliminate the need to purchase and run separate DNA and RNA purification kits, helping streamline your workflow and maximizing efficiency.
Features of the MagMAX Sequential DNA/RNA Kit include:
- Test utilization management—obtain more information using minimal sample volume to help preserve precious samples
- Versatile workflow solutions—flexible workflow enables low- to high-throughput sample processing and various sample input volume formats
- Easy identification—color-coded lid labels enable quick reagent identification, helping to minimize the risk of costly errors
- Sequential DNA and RNA isolation—obtain high-quality genomic DNA and total RNA from the same blood or bone marrow sample
- Compatible with KingFisher instruments—for a streamlined, automated workflow solution
- Expand hematologic research—separate, ready-to-use DNA and RNA eluates for use in broad range of downstream applications
- Available as stand-alone products—reagents available in larger volume bottles sold separately
Made for KingFisher purification systems
The MagMAX Sequential DNA/RNA Kit is compatible with the KingFisher Duo Prime, KingFisher Flex, and KingFisher Apex purification systems, allowing for a semi-automated workflow with a total turnaround time in about 2.5 hours. By isolating genomic DNA and total RNA from a single sample in one sequential workflow, the MagMAX Sequential DNA/RNA Kit simplifies nucleic acid purification for a broad range of molecular downstream applications, such as PCR and sequencing. This all-in-one kit is an excellent choice for researchers seeking an automated, efficient workflow that delivers reliable results from limited sample material.
Working with higher throughput sample numbers or input volumes?
The MagMAX Sequential DNA/RNA Kit reagents are also available as stand-alone products sold separately. These reagents come in larger volumes to facilitate high-throughput sample processing and accommodate higher sample input volumes. When purchased as a set of eleven stand-alone reagents, users will have sufficient MagMAX Sequential DNA/RNA reagent volumes to process 440 samples ranging from 200–500 μL each.
Specifications
Specifications
| Contenu et stockage | Conserver à température ambiante |
| Format | Reagent |
| Isolation Technology | Magnetic Bead |
| Type d’échantillon | Whole Blood, Bone Marrow |
| À utiliser avec (équipement) | KingFisher™ Duo Prime, Flex, or Apex purification systems |
| À utiliser avec (application) | Molecular applications, including qPCR and sequencing |
| Type d’acide nucléique | Genomic DNA, Total RNA, miRNA |
| Gamme de produits | MagMAX |
| Type de produit | Solution de lyse/liaison de l’ADN |
| Quantité | 80 mL |
| Show More |
Usage exclusivement réservé à la recherche. Ne pas utiliser pour des procédures de diagnostic.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.