Learn More
Sulfate de magnésium, solution aqueuse 1 M, stérile, Thermo Scientific Chemicals
Code produit 15423699
Modifier l'affichage
Click to view available options
Quantité:
125 mL
250 mL
Conditionnement:
125mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15423699 125 mL 125mL
15433699 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 15423699 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur J61030.=E
Demander un format en vrac ou personnalisé
Please to purchase this item. Need a web account? Register with us today!
CAS: 7487-88-9 | MgO4S | 120.36 g/mol
Magnesium sulfate is applied in replacement therapy for hypomagnesemia. It can be used orally as a laxative to relieve occasional constipation, and to treat low levels of magnesium. It works by replacing magnesium in the body and increasing water in the intestines. It is the first-line antiarrhythmic agent for torsades de pointes in cardiac arrest under the 2005 ECC guidelines and for managing quinidine-induced arrhythmias. It is effective in decreasing the risk that pre-eclampsia progresses to eclampsia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubilité
Entièrement miscible dans l’eau. Soluble dans l’alcool, l’éther (11,6 mg/ml à 18°C) et la glycérine. Insoluble dans l’acétone.
Remarques
Stable dans les conditions de stockage recommandées. Stocker dans des conditions fraîches et sèches et dans des récipients bien scellés.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 7487-88-9
Formule moléculaire MgO4S
Poids moléculaire (g/mol) 120.36
Numéro MDL MFCD00011110
Clé InChI CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonyme magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
CID PubChem 24083
ChEBI CHEBI:32599
Nom IUPAC magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O
Spécification
Nom chimique ou matériau Magnesium sulfate
Note relative au nom sterile
Forme physique Liquide
Quantité 125 mL
Formule linéaire MgSO4
Informations sur la solubilité Fully miscible in water. Soluble in alcohol,ether (11.6mg/ml at 18°C) and glycerin. Insoluble in acetone.
Poids de la formule 120.37
Concentration or Composition (by Analyte or Components) 1M aq. soln.