missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Malt Extract Broth
Click to view available options
Quantité:
500 g
1 kg
Conditionnement:
1kg
500g
Description
Medium was recommended by Thom and Church for the cultivation of fungi.
Medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.
Dissolve 15.0 g in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.
Ingredients:
Malt Extract Base: 6.0 g/L
Dextrose: 6.0 g/L
Maltose: 1.8g/L
Yeast: 1.2 g/L
Spécification
Spécification
| Quantité | 500 g |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu