missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAP2K4 (MEK4), GST Tag Recombinant Protein

Code produit. 17178745
Click to view available options
Quantité:
1 mg
10 μg
100 μg
Conditionnement:
100µg
10µg
1mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17178745

Marque: Invitrogen™ A32894

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This MAP2K4 (MEK4) recombinant human protein (full length) was expressed in insect cells. MAP2K4 (MEK4) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for MAP2K4 (MEK4) is NP003001.1.

MAP2K4 (MEK4) serine/threonine kinase (mitogen-activated protein kinase 4) MEK4 or MKK4 is a dual specificity kinase of the STE7 family that phosphorylates and activates JNK1 and -2 as well as p38. MAP2K4 preferentially phosphorylates the tyrosine residue of JNKs. MAP2K4 mediates a variety of physiological and pathophysiological processes such as responses to cellular stresses and inflammatory cytokines. MAP2K4 is important for carcinogenesis; a loss-off-function mutation in MAP2K4 was found in lung and pancreatic tumors. Conversely, a pro-oncogenic role was also found for MAP2K4.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P45985
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50mM tris HCl with 150mM NaCl, 0.1mM PMSF, 25% glycerol, 10mM glutathione, 0.25mM DTT, 0.1mM EDTA and no preservative; pH 7.5
Identification génétique (Entrez) 6416
Poids moléculaire 70.6 kDa
Nom Human MAP2K4 (MEK4), GST Tag
Plage de pH 7.5
Méthode de purification Purified
Quantité 100 μg
Conditions de stockage -80°C, Avoid Freeze/Thaw Cycles
État réglementaire RUO
Alias de gène C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4a; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4a; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein
Nom usuel MAP2K4
Symbole de gène(s) MAP2K4
Famille des kinases MAP Kinases
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine GST-tag
Séquence Full length
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Conditions d’expédition Dry Ice
Système d’expression Insect cells
Forme protéique Full Length, Recombinant, Active
Forme Liquid
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis