missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAPK11 (p38 beta), His Tag Recombinant Protein

Code produit. p-7181518
Click to view available options
Quantité:
10 μg
Conditionnement:
10µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 11523507

Marque: Invitrogen™ PV3679

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

MAPK11 (p38 beta) is a serine/threonine kinase which is involved in a variety of cellular processes, and is activated by proinflammatory cytokines and environmental stress. Mitogen-activated protein kinase 11. The protein encoded by p38 MAPK beta is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation, and development. This kinase is most closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and environmental stress. This kinase is activated through its phosphorylation by MAP kinase kinases (MKKs), preferably by MKK6. Transcription factor ATF2/CREB2 has been shown to be a substrate of this kinase.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q15759
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50mM tris with 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol and no preservative; pH 7.5
Identification génétique (Entrez) 5600
Poids moléculaire 47.2 kDa
Nom Human MAPK11 (p38 beta), His Tag
Plage de pH 7.5
Méthode de purification Purified
Quantité 10 μg
Conditions de stockage -80°C, Avoid Freeze/Thaw Cycles
État réglementaire RUO
Alias de gène MAP kinase 11; MAP kinase p38 beta; MAPK 11; Mapk11; mitogen activated protein kinase 11; mitogen-activated protein kinase 11; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; OTTHUMP00000196655; p38-2; P38b; p38beta; P38BETA2; Prkm11; protein kinase, mitogen activated kinase, 11, p38beta; SAPK2; SAPK2b; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b
Nom usuel p38 MAPK beta
Symbole de gène(s) MAPK11
Famille des kinases MAP Kinase Family
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine His-tag
Séquence Full length
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Entrée du dosage Biochemical terbium phospho-specific activity, Biochemical phospho-specific activity (no antibody)
Groupe de kinases Ser⁄Thr Kinases: CMGC Group
Conditions d’expédition Dry Ice
Compatibilité des systèmes LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Entrée cible MAPK11
Gène cible mitogen-activated protein kinase 11
Système d’expression Insect cells
Famille de protéines Kinases & Inhibitors
Forme protéique Recombinant, Full Length
Forme Liquid
Sous-type de protéine Serine/Threonine Kinases
Catégorie de recherche Signal Transduction
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis