Learn More
Invitrogen™ Human MAPK12 (p38 gamma), His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
Specifications
Specifications
| Numéro d’adhésion | P53778 |
| Concentration | See Label |
| À utiliser avec (application) | Kinase Assay |
| Formule | 50mM tris with 50% glycerol, 0.5mM EDTA, 0.02% Triton X-100, 150mM NaCl, 2mM DTT and no preservative; pH 7.5 |
| Identification génétique (Entrez) | 6300 |
| Poids moléculaire | 46 kDa |
| Nom | Human MAPK12 (p38 gamma), His Tag |
| Plage de pH | 7.5 |
| Méthode de purification | Purified |
| Quantité | 10 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.