Learn More
Eurofins DiscoverX MEK1 Protein, Unactive
Description
ERKs (extracellular signal-regulated kinase) are activated by the MEK (mitogen-activated protein kinase, or ERK kinase). MEKs are activated by members of the Raf family. The signaling pathway comprising Raf and ERK (extracellular signal-regulated kinase) lies downstream of the small guanine nucleotide binding protein Ras and mediates several apparently conflicting cellular responses, such as proliferation, apoptosis, growth arrest, differentiation, and senescence, depending on the duration and strength of the external stimulus and on cell type. An inhibitor assay may be of benefit for the treatment of cancer, diabetes, neurodegenerative disorders or asthma.
EMBL Z30163, possessing amino acid substitution A327G. Please note, introduction of the A327G mutation to the rabbit sequence (Z30163) results in expression of recombinant MEK1 protein that is 100% identical to wild-type human MEK1 described in SwissProt Q02750, note this SwissProt entry fails to record M1 and starts at P2.
Spécification
Spécification
| Formule | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 5% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution. |
| Poids moléculaire | 71 kDa |
| Quantité | 50 μg |
| Espèces | Human |
| Recombinant | Recombinant |
| Marqueur de protéine | N-terminal GST and C-terminal 6His tags |
| Système d’expression | Recombinant enzyme expressed in E. coli cells |
| Forme protéique | Full-length |
| Pureté ou qualité | ≥65% |
| Protéine | MEK1, unactive |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.