Learn More
STEMCELL Technologies MesenCult™ MSC Basal Medium (Human)
Click to view available options
Quantité:
450 ml
Description
- MesenCult™ MSC Basal Medium (Human) is a standardized basal medium designed to be supplemented with MesenCult™ Mesenchymal Stem Cell Stimulatory Supplement (Human; Catalog #05402) for the in vitro culture of human mesenchymal stem cells (MSCs)
- MesenCult™ MSC Basal Medium is a component of MesenCult™ Proliferation Kit (Human; Catalog #05411), and is also available separately
Spécification
Spécification
| Type de cellule | Mesenchymal Stem and Progenitor Cells |
| À utiliser avec (application) | Cell Culture, Colony Assay, Expansion |
| Quantité | 450 ml |