Learn More
Thermo Scientific Chemicals Met-Enkephalin
Code produit. p-7042039
Click to view available options
Quantité:
25 mg
50 mg
125 mg
Conditionnement:
1 pièce
25mg
50mg
Code produit. 15423789
Marque: Thermo Scientific Chemicals J61154.MB
Demander un format en vrac ou personnalisé
CAS: 58569-55-4573.665 g/mol
Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Endogenous opioid peptide that also functions as a growth factor on many cell types at a receptor that is distinct from the neuronal opioid receptors
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 58569-55-4
Spécification
Couleur White
Contenu et stockage Store at -20°C
Format Poudre
Poids moléculaire 573.665
Type de produit Met-Enkephalin
Quantité 25 mg