Learn More
Microbiologics™ Citrobacter freundii ATCC® 43864™†
Code produit. 11946725
Encompass_Preferred
Click to view available options
Quantité:
2/paquet
Type de produit:
KWIK-STIK
Code produit. 11946725
Marque: Microbiologics™ 0229P
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Citrobacter freundii strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Citrobacter freundii strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 43864™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Spécification
ATCC® ATCC 43864
Niveau de sécurité biologique 1
Certifications/Conformité CE, online certificate of analysis
Contenu et stockage 2°C à 8°C
Forme Lyophilized Pellet
Type de produit KWIK-STIK
À utiliser avec (application) Culture purpose and QC application
Quantité 2/paquet
Espèces Citrobacter freundii