Learn More
Kern Sohn Caméra de microscope ODC
Click to view available options
Type de port:
USB 2.0
USB 3.0
Résolution:
20 MP
MP de 5,1
Dimensions:
1/2,5 po Capteur
1 po Capteur
Compact, 1 po Capteur
Conditionnement:
1 pièce
Description
- The universal microscope cameras can be used anywhere and can be connected to the microscope as well as to a laptop or PC using the USB cable (USB 2.0)
- Through the proven CMOS technology, in connection with the USB 2.0 the images are shown quickly and clearly
- These cameras are also ideal for more demanding applications, such as, for example, darkfield, phase contrast and for fluorescence applications.
- Your daily work is made significantly easier with the very best synchronisation, a high frame rate as well as stable image performance together with our Microscope VIS software which we deliver with the product.
- The power supply is through the USB cable, which means that no additional power supply is required
- As well as the camera, the delivery includes our multi-lingual KERN Microscope VIS Basic KERN OXM 901, a USB cable (Length: 2000 mm) various eyepiece adapters and an object micrometer to calibrate the software.
- Please order the appropriate C-mount adapter to fit your KERN microscope now.
Spécification
Spécification
| À utiliser avec (équipement) | Microscopes |
| Type de port | USB 3.0 |
| Résolution | MP de 5,1 |
| Types de capteur | CMOS |
| Dimensions | 1/2,5 po Capteur |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.