Learn More
Invitrogen™ Mouse anti-Human IgG1 Fc Secondary Antibody, Alexa Fluor™ 488
Code produit 10794259
Modifier l'affichage
Click to view available options
Quantité:
250 μg
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
10794259 250 μg 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 10794259 Fournisseur Invitrogen™ Code fournisseur A10631
Suivez le workflow : Explorez les économies
Utilisez le code promo 27967
Afficher toutes les promotions
 Article restreint
Expédition estimée: 30-11-2026
Mouse Monoclonal Secondary Antibody
Previous lots were labeled as Mouse anti-Human IgG1. Both descriptions are correct. Please reference https://pubmed.ncbi.nlm.nih.gov/3069872/ for clone information. Product will be shipped at Room Temperature.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Human IgG1 Fc
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone HP6069
Concentration 1 mg/mL
Conjugué Alexa Fluor 488
Formule PBS with 5mM sodium azide; pH 7.5
Espèces hôtes Mouse
Immunogène IgG gamma 1.
Méthode de purification Purified
Quantité 250 μg
État réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Human
Contenu et stockage 4°C, store in dark
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG1 κ