missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse IL-10 Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
Description
Recombinant Murine IL-10 is an 18.7 kDa protein of 161 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
Spécification
Spécification
| Numéro d’adhésion | P18893 |
| À utiliser avec (application) | ELISA, Functional Assay, Western Blot |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 16153 |
| Poids moléculaire | 18.7 kDa |
| Nom | Mouse IL-10 |
| Quantité | 2 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu