missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-15/IL-15R Complex

Code produit. 15517506
Click to view available options
Quantité:
25 μg
5 μg
Conditionnement:
25µg
5µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 15517506

Marque: Invitrogen™ 14815280

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Interleukin-15 (IL-15) is a 14 kDa proinflammatory protein that has been shown to play a role in the activation of neutrophils, dendritic cells, and macrophages, and is essential to the development and survival of NK cells and CD8 T-cells. IL-15 activates Jak/Stat pathways by signaling through a heterotrimeric receptor. Two of the subunits of this receptor, beta and gamma, are shared with the IL-2 receptor, while the alpha subunit is unique to IL-15.Despite the expression of IL-15 mRNA in many cell types, the secreted protein is rarely detectable in biological samples. Recent research suggests that IL-15 is retained inside the cell and is only secreted in complex with its unique receptor, IL-15R alpha. This chaperoning is required from the ER to Golgi through to secretion. The cytokine/receptor complex may stay bound to the cell surface, where it can be trans-presented to cells expressing the beta/gamma subunits of the receptor, or may be secreted in a soluble form that lacks the transmembrane domain of the receptor. The soluble IL-15/IL-15R complex is produced by mouse dendritic cells, fibroblasts, and macrophages, and demonstrates a 10-100-fold increase in agonistic activity over IL-15 alone. Description: This recombinant protein is the heterocomplex of the mouse IL-15 linked to soluble IL-15 receptor alpha chain (sushi domain) via a GS linker. Interleukin 15 (IL-15) is a cytokine that stimulates T and NK cell proliferation and differentiation. IL-15 regulates macrophage proinflammatory and chemotactic protein production. In addition, it induces differentiation of monocytes towards dendritic cells. The receptor for IL-15 is a high affinity heterotrimeric receptor composed of IL-15Ralpha and 2 additional subunits of the IL-2 receptor (beta and gamma). Upon activation, the receptor complex signals via the Stat and JAK pathways. It has been reported that the soluble form of IL-15Ralpha corresponding to the entire extracellular domain, cleaved by MMP′s, behaves as a high affinity IL-15 antagonist, whereas the recombinant extracellular domain comprised of the sushi domain together with the ligand, IL-15, behaves as a potent agonist by enhancing binding of IL-15 to the receptor complex. In fact, this receptor-ligand complex has approximately 10-100 fold increased proliferative activity. Applications Reported: Recombinant mouse IL-15/IL-15RA complex is biologically active. Applications Tested: The ED50 of this protein, as measured by CTLL-2 cell proliferation assay, is less than or equal to 4ng/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e5 Units/mg. Source: Insect cell expressed N-terminal His-Tagged mIL-15Ra Gly33-Ala132 accession &Num; NP_032384-GS linker-mIL-15 Asn49-Ser162 accession &Num; NP_032383. Bioactivity: The ED50 of this protein, as measured by CTLL-2 cell proliferation assay, is less than or equal to 4ng/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 10e5 Units/mg. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Molecular Weight: 29.4 kDa. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2μm post-manufacturing filtered.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P48346, Q60819
Concentration 0.1mg/mL
À utiliser avec (application) Control, Functional Assay
Formule TBS with 1% BSA and no preservative; pH 8.0
Identification génétique (Entrez) 16168, 16169
Poids moléculaire 29.4kDa
Nom Mouse IL-15/IL-15R Complex
Plage de pH 8.0
Méthode de purification SDS-PAGE
Quantité 25 μg
Source Insect cells
Conditions de stockage -80°C
État réglementaire RUO
Forme structurelle Insect cell expressed N-terminal His-Tagged mIL-15Ra Gly33-Ala132 accession # NP_032384 - GS linker - mIL-15 Asn49-Ser162 accession # NP_032383
Concentration en endotoxines Less than 0.1ng/μg cytokine as determined by the LAL assay.
Nom usuel Mouse IL-15/IL-15R Complex
Symbole de gène(s) Il15, Il15ra
Activité biologique The ED50 of this protein, as measured by CTLL-2 cell proliferation assay, is less than or equal to 4ng/mL. This corresponds to a specific activity of greater than or equal to 2.5 x 105 Units/mg.
Type de produit Recombinant Protein
Réactivité spécifique Mouse
Conjugué Unconjugated
Réactivité croisée Ms
Espèces Baculovirus
Recombinant Recombinant
Forme Liquid
Sous-type de protéine Recombinant
Pureté ou qualité Affinity chromatography
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis