missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse IL-17D Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
250µg
25µg
500µg
5µg
Description
Recombinant Murine IL-17D is a 40.6 kDa, disulfide-linked homodimer of two 183 amino acid, polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.
Spécification
Spécification
| À utiliser avec (application) | ELISA, Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 239114 |
| Poids moléculaire | 40.6 kDa |
| Nom | Mouse IL-17 D |
| Quantité | 250 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <1 EU/ μg |
| Alias de gène | AI462269; FLJ30846; IL17D; IL-17 D; IL-22; IL-27; Il27a; IL-3 R; Interleukin; interleukin 17 D; interleukin 27 A; interleukin-17 D; Interleukin-27; UNQ3096/PRO21175 |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu