missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse IL-6, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantité:
1 mg
10 μg
100 μg
500 μg
Conditionnement:
100µg
10µg
1mg
500µg
Description
Recombinant Murine IL-6 is a 21.7 kDa protein containing 188 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
Spécification
Spécification
| Numéro d’adhésion | P08505 |
| À utiliser avec (application) | Functional Assay |
| Formule | protein with no preservative |
| Identification génétique (Entrez) | 16193 |
| Poids moléculaire | 21.7 kDa |
| Nom | Mouse IL-6, Animal-Free |
| Quantité | 100 μg |
| Source | E. coli |
| État réglementaire | RUO |
| Concentration en endotoxines | <0.1 EU/ μg |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu