missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse Neuropoietin Recombinant Protein, PeproTech®

Código de producto. 17890633
missing translation for 'orderingAttributeHoverText'
Quantité:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
missing translation for 'unitSize'
100µg
1mg
250µg
25µg
500µg
5µg
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17890633

missing translation for 'mfr': Gibco™ 25025B100UG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Murine Neuropoietin is a 19.8 kDa protein containing 183 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Neuropoietin is a newly identified member of the IL-6 cytokine family. Members of this family, including IL-6, IL-11, oncostatin M, leukemia inhibitory factor (LIF), cardiotrophin-1 (CT-1), cardiotrophin-like cytokine, and CNTF, display a four-helix bundle structure, and signal through gp130-containing receptor complexes. Neuropoietin, which is predominantly expressed in neuroepithelia during embryonic life, acts through a receptor complex formed of a CNTF receptor-alpha component, gp 130, and LIF receptor. Like CNTF, it promotes the survival of embryonic motor neurons, and could increase the proliferation of neural precursor cells in the presence of EGF and FGF-2. Interestingly, the human neuropoietin gene has evolved toward a pseudogene, suggesting that alternative signaling via CNTF is an effective compensatory pathway.
TRUSTED_SUSTAINABILITY

Especificaciones

Numéro d’adhésion P83714
À utiliser avec (application) Functional Assay
Formule protein with no preservative
Identification génétique (Entrez) 244218
Poids moléculaire 19.8 kDa
Nom Mouse Neuropoietin
Quantité 100 μg
Source E. coli
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène cardiotrophin 2; cardiotrophin-2; CT-2; Ctf2; CTF2P; Gm494; Neuropoietin; Np
Nom usuel Neuropoietin
Symbole de gène(s) Ctf2
Activité biologique The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is 0.5-0.8 ug/ml
Conjugué Unconjugated
Recombinant Recombinant
Séquence MAPISPSEPI GQAYSLALYM QKNTSALLQT YLQHQGSPFS DPGFSAPELQ LSTLPSAAVS FKTWHAMEDA ERLSRAQGAF LALTQHLQLV GDDQSYLNPG SPILLAQLGA ARLRAQGLLG NMAAIMTALG LPIPPEEDTL GFVPFGASAF ERKCRGYIVT REYGHWTDRA VRDLALLKAK YSA
Contenu et stockage -20°C
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.