missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse SCF Recombinant Protein
Click to view available options
Quantité:
10 μg
100 μg
25 μg
500 μg
Conditionnement:
100µg
10µg
25µg
500µg
Description
Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
Spécification
Spécification
| Numéro d’adhésion | P20826 |
| Concentration | 200 μg/mL |
| À utiliser avec (application) | Control, ELISA, Western Blot |
| Formule | 10 mM sodium phosphate with 150 mM NaCl and no preservative; pH 7.2 |
| Identification génétique (Entrez) | 17311 |
| Poids moléculaire | 18 kD |
| Nom | Mouse SCF (aa 26-189) |
| Quantité | 10 μg |
| Source | E. coli |
| Conditions de stockage | -20°C |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu