Learn More
CPC (Colder) MPCTM Quick-Disconnect Fittings, Plug
Click to view available options
Couleur:
Teinte ambrée
Teinte violette
Conditionnement:
25 pièces
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Spécification
Spécification
| Certifications/Conformité | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Température min. de fonctionnement (métrique) | -40°F |
| Port de sortie | Plug |
| Forme | Plug |
| Matériau (joint) | Platinum-cured Silicone |
| Couleur | Teinte ambrée |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.