Learn More
Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES
Click to view available options
Quantité:
100 x 5 ml
10 x 11 ml
100 x 11 ml
Conditionnement:
10 pièces
100 pièces
Description
Perform high-quality susceptibility testing with a complete range of supplies, including Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Spécification
Spécification
| Description | Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES |
| Type de produit | Microbiology Media |
| Type de test | Microbial Susceptibility |
| Nb par paquet | 100 |
| Type d’emballage | 5 mL/Bottle |
| Quantité | 100 x 5 ml |