missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MV1, MedChemExpress
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Description
MV1 is an antagonist of IAP (inhibitor of apoptosis protein), leads to protein knockdown of HaloTag-fused proteins when combined with HaloTag ligand.
Specifications
Specifications
| Nom chimique ou matériau | MV1 |
| CAS | 1001600-54-9 |
| Couleur | Blanc |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C33H44N4O5 |
| Quantité | 100 mg |
| Informations sur la solubilité | DMSO : ≥ 125 mg/mL (216.74 mM) |
| SMILES | O=C(OC)[C@H](C(C1=CC=CC=C1)C2=CC=CC=C2)NC([C@H]3N(C([C@H](C4CCCCC4)NC([C@H](C)NC)=O)=O)CCC3)=O |
| Poids moléculaire (g/mol) | 576.73 |
| Poids de la formule | 576.73 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction