Learn More
Invitrogen™ Human MYLK4, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This MYLK4 recombinant human protein (full length) was expressed in insect cells. MYLK4 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for MYLK4 is NP_001012418.2 .
Spécification
Spécification
| Numéro d’adhésion | Q86YV6 |
| Concentration | See Label |
| À utiliser avec (application) | Kinase Assay |
| Formule | 50mM tris HCl with 150mM NaCl, 0.1mM EDTA, 0.1mM PMSF, 10mM glutathione, 25% glycerol, 0.25mM DTT and no preservative; pH 7.5 |
| Identification génétique (Entrez) | 340156 |
| Poids moléculaire | 70.9 kDa |
| Nom | Human MYLK4, GST Tag |
| Plage de pH | 7.5 |
| Méthode de purification | Purified |
| Quantité | 10 μg |
| Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.