Learn More
N-(2-hydroxyéthyl)phtalimide, 99 %, Thermo Scientific Chemicals
Tous les produits Thermo Scientific Chemicals Produits
Click to view available options
Quantité:
25 g
100 g
500 g
Conditionnement:
Flacon en verre
Conditionnement:
100g
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Nom chimique ou matériau | N-(2-Hydroxyethyl)phthalimide |
| % min. CAS | 98.5 |
| % max. CAS | 100.0 |
| Point de fusion | 126°C to 129°C |
| Couleur | Blanc |
| Spectre IR | Authentique |
| Plage de pourcentage du dosage | 98.5% min. (on dry substance) (HPLC) |
| Conditionnement | Flacon en verre |
| Quantité | 25 g |
| Poids de la formule | 191.19 |
| Afficher plus |