Learn More
N,N’-Diallyl-L-tartardiamide, 99 %, Thermo Scientific Chemicals
Code produit. p-7081977
Click to view available options
Quantité:
50 g
250 g
Conditionnement:
250g
50g
Code produit. 11434853
Marque: Thermo Scientific Alfa Aesar A12195.18
Demander un format en vrac ou personnalisé
CAS: 58477-85-3 | C10H16N2O4 | 228.25 g/mol
Cross-linking co-monomer for the polymerization of soluble polyacrylamide gels
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 58477-85-3
Formule moléculaire C10H16N2O4
Poids moléculaire (g/mol) 228.25
Numéro MDL MFCD00008640
Clé InChI ZRKLEAHGBNDKHM-OCAPTIKFSA-N
Synonyme n,n'-diallyltartardiamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-enyl butanediamide, 2r,3s-2,3-dihydroxy-n,n'-bis prop-2-en-1-yl succinamide
CID PubChem 6994946
Nom IUPAC (2S,3S)-2,3-dihydroxy-N,N’-bis(prop-2-ényl)butanediamide
SMILES O[C@@H]([C@@H](O)C(=O)NCC=C)C(=O)NCC=C
Spécification
Nom chimique ou matériau N,N'-Diallyl-L-tartardiamide
Point de fusion 184°C to 186°C
Quantité 50 g
Beilstein 1712934
Rotation optique +118° (c=3 in methanol)
Poids de la formule 228.25
Pourcentage de pureté 99%