Learn More
Nano-Secondary™ Alpaca, anti-Human, CoraLite Plus 647, Chromotek
Code produit 18006865
Modifier l'affichage
Click to view available options
Quantité:
10 μL
100 μL
Conditionnement:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18006865 10 μL 10µL
18058695 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18006865 Fournisseur Chromotek Code fournisseur shuGCL647210
 Article restreint
En stock
Ajouter au panier
Ajouter au panier
Alpaca Recombinant Monoclonal Antibody
Nano-Secondary™ alpaca anti-human IgG, recombinant VHH, is a human IgG-specific (IgG1, IgG2, IgG3, IgG4) antibody. This secondary antibody product consists of a Nanobody that binds to the Fc-domain of human IgG with high affinity and specificity.
Spécification
Antigène Nano-Secondary™
Conjugué CoraLite Plus 647
Espèces hôtes Alpaca
Quantité 10 μL
Primaire ou secondaire Secondary
Espèces cibles Human
Forme Liquid
Isotype IgG