Learn More
Macherey-Nagel™ Nanocontrol Multistandard for Drinking Water
Check several parameters at once. Macherey-Nagel™ Nanocontrol Multistandard for Drinking Water has the compensation and concentration of parameters to meet the requirements of drinking water production, with the delivery and usage convenience of a single-parameter standard.
Brand: Macherey-Nagel™ 925018
Description
Nanocontrol standards enable users to check the accuracy of the Nanocolor™ system to verify the correctness of the results. The ready-to-use standard solution can be used with Nanocolor™ photometers and test kits for easy analytical quality control and simple documentation.
The standard solution has a defined concentration in the middle of the measuring range of the test kits, with a narrow confidence interval. The standard solution is used in the test instead of a normal sample. When the result of the test is within the confidence interval, it confirms that all components of the system are working correctly and that no handling error was made.
- Parameters / Nanocolor™ tests: Aluminum, Ammonium, Chloride, Iron, Manganese and Sulphate (tests 1-02, 0-98, 1-05, 1-20, 0-21, 1-36, 0-37, 1-60, 0-58 and 0-86)
- Nanocontrol Standard: Checks instruments, reagents, accessories and handling procedures (Recommended frequency: every 10th sample for each parameter, at least 1x per month)
- Nanocontrol 100+ Solution: Spiking solution to examine possible interferences from the sample (Recommended frequency: when results are not plausible or when composition of the sample changes, at least 1x per quarter)
Specifications
| Aluminum, Ammonium, Chloride, Iron, Manganese and Sulphate | |
| Drinking water analysis | |
| Multistandard and 100+ Spiking solutions, Evaluation sheet | |
| 1 Set | |
| Nanocolor™ |
| Solution | |
| Nanocolor™ photometers and tests | |
| Standard solution: 15 - 30 determinations, Spike solution: 15 determinations | |
| Concentration: 0.50mg/L Al, 0.20mg/L NH4-N, 20mg/L Cl, 1.50mg/L Fe, 1.5mg/L Mn, 120mg/L SO4 | |
| Multistandard solutions |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.