Learn More
Nickel, Ni/Cr/Co/Mo/Ti, Alloy 282, N07208, ARMI∣MBH
Code produit. 30041162
Click to view available options
Conditionnement:
1 pièce
Code produit. 30041162
Marque: ARMI/MBH IARMNI28218X
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Nickel, Ni/Cr/Co/Mo/Ti, Alloy 282, N07208, Ø38x3mm
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Boron/Zirconium/Phosphorus/Magnesium/Copper/Vanadium/Manganese/Tungsten/Silicon/Niobium/Carbon/Iron/Aluminium/Titanium/Molybdenum/Cobalt/Chromium/Nickel/Barium/Beryllium/Bromine/Cadmium/Cerium/Cesium/Chlorine/Dysprosium/Erbium/Europium/Fluorine/Gadolinium/Germanium/Gold/Hafnium/Holmium/Indium/Iodine/Iridium/Lanthanum/Lead/Lithium/Lutetium/Mercury/Osmium/Palladium/Platinum/Praseodymium/Rhenium/Rhodium/Ruthenium/Samarium/Scandium/Silver/Tellurium/Terbium/Thallium/Thorium/Thulium/Uranium/Ytterbium/Antimony/Bismuth/Neodymium/Strontium/Potassium/Rubidium/Sodium/Calcium/Oxygen/Yttrium/Arsenic/Zinc/Tantalum/Tin/Selenium/Gallium/Nitrogen
CAS 7440-02-0
Forme Disc
Concentration or Composition (by Analyte or Components) Boron 0.0014 %/Zirconium 0.0014 %/Phosphorus 0.003 %/Magnesium 0.0054 %/Copper 0.012 %/Vanadium 0.013 %/Manganese 0.042 %/Tungsten 0.043 %/Silicon 0.054 %/Niobium 0.058 %/Carbon 0.06 %/Iron 0.9 %/Aluminium 1.57 %/Titanium 2.2 %/Molybdenum 8.4 %/Cobalt 10.38 %/Chromium 19.4 %/Nickel 56.8 %/Barium (<0.0001) %/Beryllium (<0.0001) %/Bromine (<0.0001) %/Cadmium (<0.0001) %/Cerium (<0.0001) %/Cesium (<0.0001) %/Chlorine (<0.0001) %/Dysprosium (<0.0001) %/Erbium (<0.0001) %/Europium (<0.0001) %/Fluorine (<0.0001) %/Gadolinium (<0.0001) %/Germanium (<0.0001) %/Gold (<0.0001) %/Hafnium (<0.0001) %/Holmium (<0.0001) %/Indium (<0.0001) %/Iodine (<0.0001) %/Iridium (<0.0001) %/Lanthanum (<0.0001) %/Lead (<0.0001) %/Lithium (<0.0001) %/Lutetium (<0.0001) %/Mercury (<0.0001) %/Osmium (<0.0001) %/Palladium (<0.0001) %/Platinum (<0.0001) %/Praseodymium (<0.0001) %/Rhenium (<0.0001) %/Rhodium (<0.0001) %/Ruthenium (<0.0001) %/Samarium (<0.0001) %/Scandium (<0.0001) %/Silver (<0.0001) %/Tellurium (<0.0001) %/Terbium (<0.0001) %/Thallium (<0.0001) %/Thorium (<0.0001) %/Thulium (<0.0001) %/Uranium (<0.0001) %/Ytterbium (<0.0001) %/Antimony (<0.0002) %/Bismuth (<0.0002) %/Neodymium (<0.0002) %/Strontium (<0.0002) %/Potassium (<0.0005) %/Rubidium (<0.0005) %/Sodium (<0.0005) %/Calcium (<0.0020) %/Oxygen (<0.0020) %/Yttrium (<0.0020) %/Arsenic (<0.0050) %/Zinc (<0.0070) %/Tantalum (<0.0100) %/Tin (0.0003) %/Selenium (0.001) %/Gallium (0.0019) %/Nitrogen (0.0073) %