missing translation for 'onlineSavingsMsg'
Learn More

Nizatidine, MedChemExpress

Code produit. p-200101086
Encompass_Preferred
Click to view available options
Quantité:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16480868

Marque: Medchem Express HYB0310

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress Nizatidine is a potent and orally active histamine H2 receptor antagonist, can be used for the research of stomach and intestines ulcers. Nizatidine works by decreasing the secretion of gastric acid the stomach makes and prevent ulcers from coming back after they have healed in animal models.

Nizatidine is a potent and orally active histamine H2 receptor antagonist, can be used for the research of stomach and intestines ulcers. Nizatidine works by decreasing the secretion of gastric acid the stomach makes and prevent ulcers from coming back after they have healed in animal models.

Specifications

Nom chimique ou matériau Nizatidine
Forme physique Solid
CAS 76963-41-2
Couleur Jaune clair
Quantité 5 g
Notes de qualité de la pureté Research
Formule moléculaire C12H21N5O2S2
À utiliser avec (application) COVID-19-immunoregulation
Informations sur la solubilité DMSO : ≥ 50 mg/mL (150.85 mM) ∣H2O : 20 mg/mL (60.34 mM; Need ultrasonic)
SMILES CN/C(NCCSCC1=CSC(CN(C)C)=N1)=C\[N+]([O-])=O
Poids moléculaire (g/mol) 331.46
Poids de la formule 331.46
Pourcentage de pureté 99.19%
Qualité Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.