Learn More
Invitrogen™ Human NTRK3 (TRKC), His Tag Recombinant Protein
Code produit. 15468463
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
Code produit. 15468463
Marque: Invitrogen™ PR5998B
Recombinant Protein
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic Domain, Cytoplasmic
NTRK3 (TRKC) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.
TRUSTED_SUSTAINABILITY
Spécification
Numéro d’adhésion Q16288
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50 mM tris HCl with 0.05% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 4 mM DTT, 50% glycerol and no preservative; pH 7.5
Identification génétique (Entrez) 4916
Poids moléculaire 41.2 kDa
Nom Human NTRK3 (TRKC), His Tag
Quantité 100 μg
État réglementaire RUO - research use only
Alias de gène AW125844; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; gp145(trkC); GP145-TrkC; neural receptor protein-tyrosine kinase (trkC); neurotrophic receptor tyrosine kinase 3; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 3; neurotrophin-3; NT-3 growth factor receptor; NTRK3; Ntrk3_tv3; TrkC; trk-C; trkC tyrosine kinase; tyrosine kinase receptor; tyrosine kinase receptor C
Nom usuel TrkC
Famille des kinases Neurotrophic Receptor (NTRK) Kinase Family
Longueur des protéines 510-825
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine His-tag
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Système d’expression Baculovirus
Forme Liquid