missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oltipraz, MedChemExpress
Click to view available options
Quantité:
10 mg
50 mg
100 mg
200 mg
500 mg
Conditionnement:
10mg
Description
Oltipraz has an inhibitory effect on HIF-1α activation in a time-dependent manner, completely abrogating HIF-1α induction at ≥10 μM concentrations, the IC50 of Oltipraz for HIF-1α inhibition is 10 μM. Oltipraz is a potent Nrf2 activator.
Spécification
Spécification
| Nom chimique ou matériau | Oltipraz |
| Forme physique | Solid |
| CAS | 64224-21-1 |
| Couleur | Red |
| Quantité | 100 mg |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C8H6N2S3 |
| À utiliser avec (application) | Cancer-programmed cell death |
| Synonyme | RP 35972 NSC 347901 |
| Informations sur la solubilité | DMSO : 6 mg/mL (26.51 mM; Need ultrasonic and warming) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction