Learn More
Solution de test rH200, à 275 mV, Thermo Scientific Chemicals
Code produit 15461787
Modifier l'affichage
Click to view available options
Quantité:
250 ml
500 ml
Conditionnement:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
15461787 500 ml 500mL
15451787 250 ml 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 15461787 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur 042131.AP
 Article restreint
Expédition estimée: 23-11-2026
Ajouter au panier
Test solution for ORP electrodes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau ORP test solution
Conditionnement Flacon en verre plastifié
Forme physique Liquide
Numéro MDL MFCD01867801
Quantité 500 ml