Learn More
Thermo Scientific™ Oxoid™ VTEC-RPLA Toxin Detection Kit
Click to view available options
Quantité:
20 tests/kit
Conditionnement:
20 tests
Description
Thermo Scientific™ Oxoid VTEC-RPLA Toxin Detection Kit uses reverse passive latex agglutination to detect verocytotoxin VT1 and VT2 produced by E. coli cultured from food and fecal specimens.
Spécification
Spécification
| Contenu et stockage | 2°C à 8°C |
| Description | VTEC-RPLA Toxin Detection Kit |
| Quantité | 20 tests/kit |