Learn More
Bohlender™ P-MRK Set Screw
Suitable for use with P-MRK Magnetic Stirrer Heads.
Marque:  Bohlender™ C934-01
Code produit. 15800917
  • ONLINE EXCLUSIVE PRICE
    77.70€
    65.71€ / 10 pièces
    Économisez 11.99€
    15% de réduction
Il en reste null
Expédition estimée: 08-01-2026
Ajouter au panier
Description
Description
  • Manufactured from a PTFE-PEEK compound
  • For use with P-MRK stirrer heads with ground joint NS 29/32 for shaft O.D. 8 mm as well as heads with ground joint NS 29/32 or NS 45/40 for shaft O.D. 10 mm and heads with flat flange DN 40 or DN 50 for shaft O.D. 10 mm
  • FDA conformity
  • Provided in packs of 3
Spécification
Spécification
White
Magnetic Stirrer Heads
Set Screw