Learn More
R&D Systems™ Pan Caspase OPH Inhibitor Q-VD, Non-omethylated
Description
Members of the caspase gene family (cysteine proteases with aspartate specificity) play significant roles in both inflammation and apoptosis.
- The IC50 range for caspases -1, -3, -8, -9, -10, and -12 is 25 - 400 nM
- Cells that are induced to undergo apoptosis can be cultured in the presence or absence of Q-VD-OPh inhibitor
- The Caspase Inhibitor binds to the active site of activated proteases
- Treated cells can then be assayed for evidence of apoptosis inhibition by examining either whole cells or cell lysates using standard apoptosis assays
- More effective in preventing apoptosis than the widely used pan Caspase Inhibitor, ZVAD-fmk
Spécification
Spécification
| Type de produit | Inhibiteur |
| Poids de la formule | 513.77 d |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantité | 1 mg |